siRNA Details
Gene Symbol: C1orf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004303.4 | NP_001004303.3 | 17, 18 | 2381 | ||
XM_005270584.3 | XP_005270641.1 | 13, 14 | 2057 | |||
XM_011540898.1 | XP_011539200.1 | 18, 19 | 2459 | |||
XM_011540899.1 | XP_011539201.1 | 16, 17 | 2369 | |||
XM_011540904.2 | XP_011539206.1 | 15, 16 | 1206 | |||
XM_011540906.2 | XP_011539208.1 | 13, 14 | 974 | |||
XM_017000544.1 | XP_016856033.1 | 18, 19 | 2145 | |||
XM_017000545.1 | XP_016856034.1 | 18, 19 | 2205 | |||
GenBank mRNA | AK093468.1 | - | 11, 12 | 892 | ||
BX648439.1 | - | 17, 18 | 2057 |
