siRNA Details
Gene Symbol: PLEKHG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004330.2 | NP_001004330.1 | 11 | 1207 | ||
GenBank mRNA | AK128530.1 | - | 11 | 1161 | ||
BC137086.1 | - | 10 | 1088 | 9021083 |
