siRNA Details
Gene Symbol: PCLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014510.2 | NP_055325.2 | 7 | 12687 | ||
NM_033026.5 | NP_149015.2 | 7 | 12687 | |||
XM_017012006.1 | XP_016867495.1 | 5 | 6867 | |||
XM_017012007.1 | XP_016867496.1 | 5 | 6851 | |||
GenBank mRNA | AB011131.1 | - | 1 | 1231 | ||
AK125999.1 | - | 2 | 2044 | |||
BX649041.1 | - | 2 | 2560 | |||
CR749552.1 | - | 2 | 1968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
