siRNA Details
Gene Symbol: Gnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212500.3 | NP_997665.1 | 8 | 1049 | ||
GenBank mRNA | BC083888.1 | - | 5 | 542 | ||
BC129081.1 | - | 8 | 954 | 7442774 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212500.3 | NP_997665.1 | 8 | 1049 | ||
GenBank mRNA | BC083888.1 | - | 5 | 542 | ||
BC129081.1 | - | 8 | 954 | 7442774 |