siRNA Details
Gene Symbol: NUTM2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_075100.1 | 7 | 3737 | |||
GenBank mRNA | AB095940.1 | - | 7 | 3737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
