siRNA Details
Gene Symbol: NUTM2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_075100.1 | 5 | 2029 | |||
GenBank mRNA | AB095940.1 | - | 5 | 2029 |
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099338.1 | NP_001092808.1 | 5 | 2082 | ||
XM_011540122.2 | XP_011538424.1 | 7 | 4925 | |||
XM_017016607.1 | XP_016872096.1 | 7 | 4926 | |||
GenBank mRNA | BC092519.1 | - | 5 | 2078 | ||
EG328792.1 | - | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
