siRNA Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017677.3 | NP_060147.2 | 3 | 374 | ||
GenBank mRNA | AK000133.1 | - | 1 | 33 | ||
AK225231.1 | - | 1 | 33 | |||
BC012399.1 | - | 3 | 352 | 3864128 | ||
DA691850.1 | - | 3 | 374 |
