siRNA Details
Gene Symbol: RGD1311899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009630.1 | NP_001009630.1 | 6 | 1072 | ||
XM_006249433.2 | XP_006249495.1 | 7 | 758 | |||
GenBank mRNA | BC088751.1 | - | 6 | 1072 | 7317722 |
