siRNA Details
Gene Symbol: Mgme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009655.1 | NP_001009655.1 | 5 | 1851 | ||
XM_006235120.2 | XP_006235182.1 | 5 | 1900 | |||
XM_006235121.1 | XP_006235183.1 | 4 | 1784 | |||
XM_006235122.2 | XP_006235184.1 | 5 | 1917 | |||
XM_006235123.2 | XP_006235185.1 | 5 | 1976 | |||
GenBank mRNA | BC087676.1 | - | 5 | 1851 | 7319345 |
