siRNA Details
Gene Symbol: Hat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009657.1 | NP_001009657.1 | 6 | 625 | ||
GenBank mRNA | BC087663.1 | - | 6 | 625 | 7315259 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026115, AK010715, AK137648, AK014330, AK169315, and others.
Gene Ontology Categories:
