siRNA Details
Gene Symbol: Wbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763375.1 | 12 | 2309 | |||
GenBank mRNA | BC087082.1 | - | 12 | 2222 | 7189215 |
Gene Symbol: Wbp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763449.1 | XP_008761671.1 | 2309 | |||
XM_008763450.1 | XP_008761672.1 | 2311 |

More Information
Other Species Targeted

Gene Ontology Categories:
