siRNA Details
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009663.1 | NP_001009663.1 | 3 | 709 | ||
GenBank mRNA | BC088300.1 | - | 3 | 709 | 7323342 | |
FQ211289.1 | - | 3 | 724 | |||
FQ218398.1 | - | 3 | 718 |

More Information
Other Species Targeted

Gene Ontology Categories:
