siRNA Details
Gene Symbol: Fastkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009673.1 | NP_001009673.1 | 3 | 958 | ||
XM_006245050.1 | XP_006245112.1 | 4 | 1085 | |||
GenBank mRNA | BC088416.1 | - | 3 | 958 | 7112913 |
