siRNA Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011890.1 | NP_001011890.1 | 2 | 210 | ||
XM_006245931.2 | XP_006245993.1 | 2 | 654 | |||
XM_006245932.2 | XP_006245994.1 | 3 | 353 | |||
GenBank mRNA | BC079014.1 | - | 2 | 210 | 7113485 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009070, NM_001080127, CJ058744, AK178555, BC002061, and others.
Gene Ontology Categories:
