siRNA Details
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011896.1 | NP_001011896.1 | 20 | 2004 | ||
GenBank mRNA | BC082042.1 | - | 20 | 2004 | 7131554 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013852, AK030626, AK003527, AF213381, BC032923.
Gene Ontology Categories:
