siRNA Details
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011905.1 | NP_001011905.1 | 22 | 2200 | ||
XM_006240747.2 | XP_006240809.1 | 22 | 2511 | |||
XM_006240748.2 | XP_006240810.1 | 22 | 2457 | |||
GenBank mRNA | BC083867.1 | - | 22 | 2200 | 7127907 |
