siRNA Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011927.1 | NP_001011927.1 | 4, 5 | 636 | ||
GenBank mRNA | BC088462.1 | - | 4, 5 | 636 | 7127604 | |
FQ213716.1 | - | 4, 5 | 652 | |||
FQ215412.1 | - | 3, 4 | 371 |
