siRNA Details
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011943.1 | NP_001011943.1 | 5 | 558 | ||
XM_006231472.1 | XP_006231534.1 | 5 | 669 | |||
XM_006231474.2 | XP_006231536.1 | 5 | 670 | |||
XM_006231475.2 | XP_006231537.1 | 3 | 3001 | |||
GenBank mRNA | BC079096.1 | - | 5 | 558 | 7124109 |
