siRNA Details
Gene Symbol: Ccna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011949.1 | NP_001011949.1 | 2 | 394 | ||
XM_006232359.2 | XP_006232421.1 | 2 | 356 | |||
XM_006232360.2 | XP_006232422.1 | 2 | 359 | |||
XM_006232361.2 | XP_006232423.1 | 2 | 399 | |||
XM_006232362.2 | XP_006232424.1 | 2 | 360 | |||
XM_006232363.2 | XP_006232425.1 | 2 | 359 | |||
XM_006232364.2 | XP_006232426.1 | 2 | 361 | |||
XM_008760987.1 | XP_008759209.1 | 2 | 449 | |||
XM_008760988.1 | XP_008759210.1 | 2 | 340 | |||
GenBank mRNA | BC079234.1 | - | 2 | 394 | 7132591 |
