siRNA Details
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011963.1 | NP_001011963.1 | 6 | 1423 | ||
XM_006233912.2 | XP_006233974.1 | 6 | 1391 | |||
XM_008761641.1 | XP_008759863.1 | 5 | 2157 | |||
GenBank mRNA | BC079099.1 | - | 6 | 1423 | 7124433 |
