siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011988.1 | NP_001011988.1 | 7, 8 | 931 | ||
XM_006241363.1 | XP_006241425.1 | 9, 10 | 1007 | |||
XM_006241364.2 | XP_006241426.1 | 9, 10 | 1080 | |||
XM_006241365.2 | XP_006241427.1 | 10, 11 | 1303 | |||
XM_006241366.2 | XP_006241428.1 | 10, 11 | 1308 | |||
XM_006241367.2 | XP_006241429.1 | 10, 11 | 1138 | |||
XM_006241369.1 | XP_006241431.1 | 8, 9 | 884 | |||
XM_006241370.2 | XP_006241432.1 | 8, 9 | 1013 | |||
GenBank mRNA | BC087653.1 | - | 7, 8 | 931 | 7313082 |
