siRNA Details
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012009.1 | NP_001012009.1 | 3 | 362 | ||
XM_006246958.1 | XP_006247020.1 | 4 | 498 | |||
XM_006246959.1 | XP_006247021.1 | 3 | 686 | |||
GenBank mRNA | BC088249.1 | - | 3 | 362 | 7370364 |
