siRNA Details
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012011.1 | NP_001012011.1 | 2 | 199 | ||
XM_006246989.2 | XP_006247051.1 | 2 | 233 | |||
XM_006246990.2 | XP_006247052.1 | 2 | 219 | |||
XM_008768020.1 | XP_008766242.1 | 1 | 1105 | |||
XM_008768021.1 | XP_008766243.1 | 1 | 1105 | |||
XM_008768022.1 | XP_008766244.1 | 1 | 1105 | |||
XM_008768023.1 | XP_008766245.1 | 2 | 233 | |||
GenBank mRNA | BC085959.1 | - | 2 | 199 | 7135421 |

More Information
Other Species Targeted

Gene Ontology Categories:
