siRNA Details
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012013.1 | NP_001012013.1 | 4 | 540 | ||
XM_006247508.2 | XP_006247570.1 | 5 | 528 | |||
XM_006247509.2 | XP_006247571.1 | 5 | 530 | |||
XM_006247510.2 | XP_006247572.1 | 5 | 529 | |||
XM_006247511.2 | XP_006247573.1 | 5 | 528 | |||
XM_006247512.2 | XP_006247574.1 | 5 | 529 | |||
XM_008768284.1 | XP_008766506.1 | 6 | 631 | |||
XM_008768285.1 | XP_008766507.1 | 5 | 519 | |||
XM_008768286.1 | XP_008766508.1 | 5 | 530 | |||
XM_008768287.1 | XP_008766509.1 | 5 | 529 | |||
GenBank mRNA | BC076387.1 | - | 4 | 540 | 7109482 |
