siRNA Details
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017752.2 | NP_060222.2 | 18 | 2924 | ||
XM_006724669.1 | XP_006724732.1 | 17 | 2774 | |||
XM_006724670.3 | XP_006724733.1 | 18 | 2738 | |||
GenBank mRNA | AB449890.1 | - | 18 | 2759 | ||
AK000305.1 | - | 3 | 345 | |||
AK225310.1 | - | 3 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
