siRNA Details
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013062.1 | NP_001013080.1 | 19 | 3449 | ||
GenBank mRNA | AF309630.1 | - | 19 | 3449 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013062.1 | NP_001013080.1 | 19 | 3449 | ||
GenBank mRNA | AF309630.1 | - | 19 | 3449 |