siRNA Details
Gene Symbol: Tspan4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013070.1 | NP_001013088.1 | 622 | |||
XM_006230545.2 | XP_006230607.1 | 844 | ||||
XM_008759985.1 | XP_008758207.1 | 740 | ||||
XM_008759986.1 | XP_008758208.1 | 547 | ||||
GenBank mRNA | BC091174.1 | - | 622 | 7321737 | ||
FQ222408.1 | - | 587 |
Gene Symbol: LOC100911766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749018.3 | XP_003749066.1 | 5 | 617 | ||
XM_003749019.3 | XP_003749067.1 | 5 | 844 | |||
XM_008760032.1 | XP_008758254.1 | 4 | 740 | |||
XM_008760033.1 | XP_008758255.1 | 5 | 547 |
