siRNA Details
Gene Symbol: DCHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017639.3 | NP_060109.2 | 24 | 6110 | ||
XM_011532045.1 | XP_011530347.1 | 19 | 7898 | |||
XM_011532046.2 | XP_011530348.1 | 18 | 5554 | |||
XM_011532049.2 | XP_011530351.1 | 11 | 3404 | |||
GenBank mRNA | AK000054.1 | - | 2 | 407 | ||
AK095103.1 | - | 7 | 1447 | |||
AK225200.1 | - | 2 | 407 | |||
AY354497.1 | - | 24 | 6110 | |||
BX647835.1 | - | 7 | 1086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
