siRNA Details
Gene Symbol: Hibch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013112.1 | NP_001013130.1 | 12 | 1017 | ||
XM_006244895.1 | XP_006244957.1 | 14 | 1257 | |||
GenBank mRNA | BC083737.1 | - | 12 | 1017 | 7191666 |

More Information
Other Species Targeted

Gene Ontology Categories:
