siRNA Details
Gene Symbol: LOC288521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013868.1 | NP_001013890.1 | 1400 | |||
GenBank mRNA | BC079301.1 | - | 1400 | 7134538 |
Gene Symbol: MGC112692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025759.1 | NP_001020930.1 | 1399 | |||
GenBank mRNA | BC081909.1 | - | 1396 | |||
BC098706.1 | - | 1399 | 7114401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
