siRNA Details
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013869.1 | NP_001013891.1 | 23 | 2474 | ||
XM_006255233.2 | XP_006255295.1 | 24 | 2666 | |||
XM_006255234.2 | XP_006255296.1 | 24 | 2663 | |||
XM_008772364.1 | XP_008770586.1 | 23 | 2532 | |||
GenBank mRNA | BC081948.1 | - | 23 | 2474 | 7122187 |
