siRNA Details
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013881.2 | NP_001013903.2 | 11, 12 | 2102 | ||
XM_006252879.2 | XP_006252941.1 | 10, 11 | 1782 | |||
XM_006252880.2 | XP_006252942.1 | 10, 11 | 1779 | |||
XM_006252881.2 | XP_006252943.1 | 10, 11 | 1773 | |||
XM_006252882.2 | XP_006252944.1 | 10, 11 | 1770 | |||
XM_006252884.2 | XP_006252946.1 | 11, 12 | 2061 | |||
XM_006252887.2 | XP_006252949.1 | 10, 11 | 1594 | |||
XM_008771089.1 | XP_008769311.1 | 12, 13 | 2151 | |||
XM_008771090.1 | XP_008769312.1 | 11, 12 | 1781 | |||
GenBank mRNA | BC089902.1 | - | 8, 9 | 1192 | 7383529 |
