siRNA Details
Gene Symbol: RGD1307537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013885.2 | NP_001013907.1 | 3 | 616 | ||
GenBank mRNA | BC079269.1 | - | 617 | 7133712 | ||
CK596086.1 | - | 3 | 616 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1307537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013885.2 | NP_001013907.1 | 3 | 616 | ||
GenBank mRNA | BC079269.1 | - | 617 | 7133712 | ||
CK596086.1 | - | 3 | 616 |