siRNA Details
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013887.1 | NP_001013909.1 | 7 | 842 | ||
XM_006254546.2 | XP_006254608.1 | 6 | 821 | |||
GenBank mRNA | BC079289.1 | - | 7 | 842 | 7134364 | |
DQ158861.1 | - | 6 | 775 | |||
FQ214241.1 | - | 6 | 695 |
