siRNA Details
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013910.2 | NP_001013932.1 | 4 | 543 | ||
GenBank mRNA | AY552804.1 | - | 4 | 470 | ||
BC097370.1 | - | 4 | 543 | 7444022 | ||
DQ120489.1 | - | 4 | 435 | |||
DQ120490.1 | - | 4 | 435 |
