siRNA Details
Gene Symbol: Ankrd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013948.1 | NP_001013970.1 | 5 | 695 | ||
XM_006241530.2 | XP_006241592.1 | 5 | 966 | |||
XM_006241531.2 | XP_006241593.1 | 4 | 598 | |||
XM_006241532.2 | XP_006241594.1 | 4 | 2339 | |||
XM_006241533.2 | XP_006241595.1 | 4 | 2265 | |||
GenBank mRNA | AB095364.1 | - | 5 | 695 | ||
BC105616.1 | - | 5 | 691 | 7453107 |
