siRNA Details
Gene Symbol: Cuedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013971.1 | NP_001013993.1 | 1 | 545 | ||
GenBank mRNA | BC087087.1 | - | 1 | 545 | 7189849 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cuedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013971.1 | NP_001013993.1 | 1 | 545 | ||
GenBank mRNA | BC087087.1 | - | 1 | 545 | 7189849 |