siRNA Details
Gene Symbol: NHLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000154.1 | - | 1 | 428 | ||
AK126751.1 | - | 1 | 1475 | |||
BC013743.1 | - | 1 | 892 | |||
BX647663.1 | - | 1 | 1980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
