siRNA Details
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001218.4 | NP_001209.1 | 11 | 3495 | ||
NM_001293642.1 | NP_001280571.1 | 9 | 3282 | |||
NM_206925.2 | NP_996808.1 | 10 | 3462 | |||
NR_135511.1 | 10 | 3565 | ||||
GenBank mRNA | AK000158.1 | - | 1 | 211 | ||
BC001012.2 | - | 1 | 352 | |||
BC087838.1 | - | 1 | 919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
