siRNA Details
Gene Symbol: Ptgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015009.1 | NP_001015009.1 | 8 | 1639 | ||
XM_006240326.1 | XP_006240388.1 | 10 | 1863 | |||
XM_006240327.1 | XP_006240389.1 | 10 | 1799 | |||
XM_006240328.2 | XP_006240390.1 | 10 | 1761 | |||
XM_006240329.2 | XP_006240391.1 | 8 | 2386 | |||
GenBank mRNA | BC091173.1 | - | 8 | 1639 | 7189599 |
