siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017448.1 | NP_001017448.1 | 2 | 198 | ||
XM_006256562.2 | XP_006256624.1 | 2 | 479 | |||
XM_006256563.2 | XP_006256625.1 | 2 | 482 | |||
XM_006256564.2 | XP_006256626.1 | 2 | 234 | |||
XM_006256565.1 | XP_006256627.1 | 3 | 257 | |||
XM_006256566.2 | XP_006256628.1 | 2 | 891 | |||
XM_006256567.2 | XP_006256629.1 | 3 | 226 | |||
XM_006256568.2 | XP_006256630.1 | 1 | 109 | |||
XM_008772982.1 | XP_008771204.1 | 2 | 1149 | |||
XM_008772983.1 | XP_008771205.1 | 2 | 222 | |||
GenBank mRNA | BC079256.1 | - | 2 | 198 | 7133287 |
