siRNA Details
Gene Symbol: MCM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017696.2 | NP_060166.2 | 12 | 3137 | ||
GenBank mRNA | AK000177.1 | - | 1 | 530 | ||
AK225233.1 | - | 1 | 530 | |||
AK299076.1 | - | 7 | 2009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
