siRNA Details
Gene Symbol: Fat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080814.1 | NP_001074283.1 | 25 | 17087 | ||
GenBank mRNA | AK039309.1 | - | 1 | 1211 | ||
AK052501.1 | - | 1 | 2825 | |||
BC030299.1 | - | 1 | 3842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
