siRNA Details
Gene Symbol: Celsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242167.2 | XP_006242229.1 | 1 | 3802 | ||
XM_008765820.1 | XP_008764042.1 | 1 | 3803 | |||
XM_008765821.1 | XP_008764043.1 | 1 | 3802 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
