siRNA Details
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268469.2 | XP_005268526.1 | 8 | 1552 | ||
XM_017009600.1 | XP_016865089.1 | 8 | 2917 | |||
GenBank mRNA | AK000202.1 | - | 4 | 918 | ||
AK056861.1 | - | 6 | 1651 | |||
AK122862.1 | - | 6 | 2987 | |||
BC002482.2 | - | 7 | 1205 |
Gene Symbol: LOC105376839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JQ358763.1 | - | 1 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
