siRNA Details
Gene Symbol: Tmem5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024759.1 | NP_001019930.1 | 5 | 938 | ||
XM_006241397.2 | XP_006241459.1 | 5 | 1003 | |||
XM_008765391.1 | XP_008763613.1 | 4 | 690 | |||
GenBank mRNA | BC097355.1 | - | 5 | 938 | 7441970 |
