siRNA Details
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024765.1 | NP_001019936.1 | 23 | 3028 | ||
XM_006247321.2 | XP_006247383.1 | 23 | 3310 | |||
XM_006247322.2 | XP_006247384.1 | 23 | 3225 | |||
XM_006247323.2 | XP_006247385.1 | 23 | 3082 | |||
XM_006247324.2 | XP_006247386.1 | 23 | 3029 | |||
XM_006247325.2 | XP_006247387.1 | 22 | 3074 | |||
XM_006247326.2 | XP_006247388.1 | 22 | 3074 | |||
GenBank mRNA | BC095903.1 | - | 23 | 3028 | 7096697 | |
BC103645.1 | - | 3 | 203 | |||
FQ211883.1 | - | 23 | 3028 |
