siRNA Details
Gene Symbol: Rogdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024864.1 | NP_001020035.1 | 6 | 596 | ||
GenBank mRNA | BC083789.1 | - | 4 | 1027 | ||
BC098001.1 | - | 6 | 596 | 7453517 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133185, BC006914, AY036117, AK002523, AK046477, and others.
Gene Ontology Categories:
