siRNA Details
Gene Symbol: Cwc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024987.1 | NP_001020158.1 | 7 | 730 | ||
XM_006242520.2 | XP_006242582.1 | 7 | 761 | |||
GenBank mRNA | BC091396.1 | - | 7 | 730 | 7323256 | |
FQ224752.1 | - | 7 | 700 |
