siRNA Details
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303270.1 | NP_001290199.1 | 9 | 1378 | ||
XM_011523663.1 | XP_011521965.1 | 9 | 1374 | |||
XM_011523664.2 | XP_011521966.1 | 10 | 1523 | |||
XM_011523666.1 | XP_011521968.1 | 9 | 1374 | |||
XM_011523670.2 | XP_011521972.1 | 9 | 1374 | |||
XM_017024219.1 | XP_016879708.1 | 9 | 1374 | |||
GenBank mRNA | AK292607.1 | - | 9 | 1374 | ||
BC034225.1 | - | 9 | 1377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
